Assay Details
| Gene Symbol: LOC4346035 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793223.2 | XP_015648709.1 | 1 | 771 | 158 | |
| GenBank mRNA | AK108596.1 | - | 1 | 746 | 158 | |
More Information
Set Membership: |
|

LOC4346035
Genomic Map