Assay Details
| Gene Symbol: LOC4351682 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762871.2 | XP_015618357.1 | 1 - 2 | 235 | 62 | |
| GenBank mRNA | AK072557.1 | - | 1 - 2 | 105 | 62 | |
More Information
Set Membership: |
|

LOC4351682
Genomic Map