Assay Details
| Gene Symbol: LOC4351682 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762871.2 | XP_015618357.1 | 5 - 6 | 740 | 104 | |
| XM_015762872.2 | XP_015618358.1 | 6 - 7 | 806 | 104 | ||
| XM_015762873.2 | XP_015618359.1 | 6 - 7 | 826 | 104 | ||
| GenBank mRNA | AK072557.1 | - | 5 - 6 | 610 | 104 | |
More Information
Set Membership: |
|

LOC4351682
Genomic Map