Assay Details
| Gene Symbol: LOC4351682 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762871.2 | XP_015618357.1 | 6 - 7 | 799 | 61 | |
| XM_015762872.2 | XP_015618358.1 | 7 - 8 | 865 | 61 | ||
| XM_015762873.2 | XP_015618359.1 | 7 - 8 | 885 | 61 | ||
| GenBank mRNA | AK072557.1 | - | 6 - 7 | 669 | 61 | |
More Information
Set Membership: |
|

LOC4351682
Genomic Map