Assay Details
| Gene Symbol: LOC4351682 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762871.2 | XP_015618357.1 | 7 - 8 | 1042 | 86 | |
| XM_015762872.2 | XP_015618358.1 | 8 - 9 | 1108 | 86 | ||
| XM_015762873.2 | XP_015618359.1 | 8 - 9 | 1128 | 86 | ||
| GenBank mRNA | AK072557.1 | - | 7 - 8 | 912 | 86 | |
More Information
Set Membership: |
|

LOC4351682
Genomic Map