Assay Details
| Gene Symbol: LOC4351682 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762871.2 | XP_015618357.1 | 8 - 9 | 1227 | 90 | |
| XM_015762872.2 | XP_015618358.1 | 9 - 10 | 1293 | 90 | ||
| XM_015762873.2 | XP_015618359.1 | 9 - 10 | 1313 | 90 | ||
| GenBank mRNA | AK072557.1 | - | 8 - 9 | 1097 | 90 | |
More Information
Set Membership: |
|

LOC4351682
Genomic Map