Assay Details
| Gene Symbol: LOC4352505 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765151.2 | XP_015620637.1 | 5 - 6 | 1501 | 82 | |
| GenBank mRNA | AK066737.1 | - | 3 - 4 | 1004 | 82 | |
| AK072241.1 | - | 5 - 6 | 1497 | 82 | ||
| DQ450958.1 | - | 3 - 4 | 384 | 82 | ||
| EU513003.1 | - | 5 - 6 | 1431 | 82 | ||
| Gene Symbol: LOC4352509 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763380.2 | 6 - 7 | 1242 | 82 | ||
More Information
Set Membership: |
|

LOC4352505
Genomic Map