Assay Details
| Gene Symbol: LOC4352553 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765060.1 | XP_015620546.1 | 4 - 5 | 1037 | 106 | |
| GenBank mRNA | AK059954.1 | - | 5 - 6 | 728 | 106 | |
| AK060201.1 | - | 4 - 5 | 1037 | 106 | ||
| AK070277.1 | - | 4 - 5 | 963 | 106 | ||
| AK071697.1 | - | 4 - 5 | 963 | 106 | ||
More Information
Set Membership: |
|

LOC4352553
Genomic Map