Assay Details
| Gene Symbol: LOC4352734 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762739.2 | XP_015618225.1 | 6 - 7 | 817 | 67 | |
| GenBank mRNA | AK072382.1 | - | 6 - 7 | 668 | 67 | |
More Information
Set Membership: |
|

LOC4352734
Genomic Map