Assay Details
| Gene Symbol: LOC4338654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784230.2 | XP_015639716.1 | 4 - 5 | 707 | 92 | |
| GenBank mRNA | AK069987.1 | - | 4 - 5 | 713 | 92 | |
More Information
Set Membership: |
|

LOC4338654
Genomic Map