Assay Details
| Gene Symbol: LOC4341640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785903.2 | XP_015641389.1 | 9 - 10 | 892 | 77 | |
| GenBank mRNA | AK065376.1 | - | 9 - 10 | 840 | 77 | |
More Information
Set Membership: |
|

LOC4341640
Genomic Map