Assay Details
| Gene Symbol: LOC4342744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790013.2 | XP_015645499.1 | 5 - 6 | 418 | 93 | |
| GenBank mRNA | AK241980.1 | - | 7 - 8 | 490 | 93 | |
More Information
Set Membership: |
|

LOC4342744
Genomic Map