Assay Details
| Gene Symbol: LOC4342744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790013.2 | XP_015645499.1 | 7 - 8 | 523 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4342744| Gene Symbol: LOC4342744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790013.2 | XP_015645499.1 | 7 - 8 | 523 | 90 | |
Set Membership: |
|