Assay Details
| Gene Symbol: LOC4344386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791030.1 | XP_015646516.1 | 10 - 11 | 1546 | 111 | |
| GenBank mRNA | AK064333.1 | - | 10 - 11 | 1531 | 111 | |
| AK102007.1 | - | 10 - 11 | 1549 | 111 | ||
More Information
Set Membership: |
|

LOC4344386
Genomic Map