Assay Details
| Gene Symbol: LOC4344858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795219.2 | XP_015650705.1 | 1 - 2 | 153 | 77 | |
| GenBank mRNA | AK066092.1 | - | 1 - 2 | 119 | 77 | |
| AK103694.1 | - | 1 - 2 | 105 | 77 | ||
| AK104866.1 | - | 1 - 2 | 71 | 77 | ||
| EF122487.1 | - | 1 - 2 | 129 | 77 | ||
| GQ848063.1 | - | 1 - 2 | 77 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015795219.2 transcripts.
Set Membership: |
|

LOC4344858
Genomic Map