Assay Details
| Gene Symbol: LOC4344858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795219.2 | XP_015650705.1 | 2 - 3 | 272 | 90 | |
| GenBank mRNA | AB017042.1 | - | 2 - 3 | 137 | 90 | |
| AK066092.1 | - | 2 - 3 | 238 | 90 | ||
| AK103694.1 | - | 2 - 3 | 224 | 90 | ||
| AK104866.1 | - | 2 - 3 | 190 | 90 | ||
| AK121126.1 | - | 1 - 2 | 1055 | 90 | ||
| EF122487.1 | - | 2 - 3 | 248 | 90 | ||
| GQ848063.1 | - | 2 - 3 | 196 | 90 | ||
More Information
Set Membership: |
|

LOC4344858
Genomic Map