Assay Details
| Gene Symbol: LOC4344858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795219.2 | XP_015650705.1 | 3 - 4 | 396 | 71 | |
| GenBank mRNA | AB017042.1 | - | 3 - 4 | 261 | 71 | |
| AK066092.1 | - | 3 - 4 | 362 | 71 | ||
| AK103694.1 | - | 3 - 4 | 348 | 71 | ||
| AK121126.1 | - | 2 - 3 | 1179 | 71 | ||
| EF122487.1 | - | 3 - 4 | 372 | 71 | ||
| GQ848063.1 | - | 3 - 4 | 320 | 71 | ||
More Information
Set Membership: |
|

LOC4344858
Genomic Map