Assay Details
| Gene Symbol: LOC4344858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795219.2 | XP_015650705.1 | 5 - 6 | 687 | 67 | |
| GenBank mRNA | AB017042.1 | - | 5 - 6 | 552 | 67 | |
| AK066092.1 | - | 5 - 6 | 653 | 67 | ||
| AK103694.1 | - | 5 - 6 | 639 | 67 | ||
| AK104866.1 | - | 5 - 6 | 605 | 67 | ||
| EF122487.1 | - | 5 - 6 | 663 | 67 | ||
| GQ848063.1 | - | 5 - 6 | 611 | 67 | ||
More Information
Set Membership: |
|

LOC4344858
Genomic Map