Assay Details
| Gene Symbol: LOC4344858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795219.2 | XP_015650705.1 | 6 - 7 | 737 | 128 | |
| GenBank mRNA | AB017042.1 | - | 6 - 7 | 602 | 128 | |
| AK066092.1 | - | 6 - 7 | 703 | 128 | ||
| AK103694.1 | - | 6 - 7 | 689 | 128 | ||
| AK104866.1 | - | 6 - 7 | 655 | 128 | ||
| EF122487.1 | - | 6 - 7 | 713 | 128 | ||
| GQ848063.1 | - | 6 - 7 | 661 | 128 | ||
More Information
Set Membership: |
|

LOC4344858
Genomic Map