Assay Details
| Gene Symbol: LOC4344858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795219.2 | XP_015650705.1 | 7 - 8 | 842 | 127 | |
| GenBank mRNA | AB017042.1 | - | 7 - 8 | 707 | 127 | |
| AK066092.1 | - | 7 - 8 | 808 | 127 | ||
| AK103694.1 | - | 7 - 8 | 794 | 127 | ||
| AK104866.1 | - | 7 - 8 | 760 | 127 | ||
| EF122487.1 | - | 7 - 8 | 818 | 127 | ||
| GQ848063.1 | - | 7 - 8 | 766 | 127 | ||
More Information
Set Membership: |
|

LOC4344858
Genomic Map