Assay Details
| Gene Symbol: LOC4344858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795219.2 | XP_015650705.1 | 8 - 9 | 994 | 64 | |
| GenBank mRNA | AB017042.1 | - | 8 - 9 | 859 | 64 | |
| AK066092.1 | - | 8 - 9 | 960 | 64 | ||
| AK103694.1 | - | 8 - 9 | 946 | 64 | ||
| AK104866.1 | - | 8 - 9 | 912 | 64 | ||
| EF122487.1 | - | 8 - 9 | 970 | 64 | ||
| GQ848063.1 | - | 8 - 9 | 918 | 64 | ||
More Information
Set Membership: |
|

LOC4344858
Genomic Map