Assay Details
| Gene Symbol: LOC4345687 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793027.2 | XP_015648513.1 | 1 | 1798 | 79 | |
| GenBank mRNA | AK108225.1 | - | 1 | 1051 | 79 | |
More Information
Set Membership: |
|

LOC4345687
Genomic Map