Assay Details
| Gene Symbol: LOC4347016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756325.2 | XP_015611811.1 | 11 - 12 | 2281 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4347016| Gene Symbol: LOC4347016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756325.2 | XP_015611811.1 | 11 - 12 | 2281 | 98 | |
Set Membership: |
|