Assay Details
| Gene Symbol: LOC4347016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756325.2 | XP_015611811.1 | 1 - 2 | 884 | 125 | |
| GenBank mRNA | AK242913.1 | - | 1 - 2 | 832 | 125 | |
More Information
Set Membership: |
|

LOC4347016
Genomic Map