Assay Details
| Gene Symbol: LOC4347016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756325.2 | XP_015611811.1 | 6 - 7 | 1280 | 94 | |
| GenBank mRNA | AK242913.1 | - | 6 - 7 | 1228 | 94 | |
More Information
Set Membership: |
|

LOC4347016
Genomic Map