Assay Details
| Gene Symbol: LOC4347277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755488.2 | XP_015610974.1 | 1 | 1218 | 104 | |
| GenBank mRNA | AB118006.1 | - | 1 | 356 | 104 | |
More Information
Set Membership: |
|

LOC4347277
Genomic Map