Assay Details
| Gene Symbol: LOC4348153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758888.2 | XP_015614374.1 | 3 - 4 | 720 | 86 | |
| GenBank mRNA | AK111889.1 | - | 3 - 4 | 673 | 86 | |
More Information
Set Membership: |
|

LOC4348153
Genomic Map