Assay Details
| Gene Symbol: LOC4348496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759277.2 | XP_015614763.1 | 6 - 7 | 1103 | 126 | |
| XM_015759278.2 | XP_015614764.1 | 6 - 7 | 1103 | 126 | ||
| GenBank mRNA | AK066800.1 | - | 6 - 7 | 1101 | 126 | |
| AK099380.1 | - | 6 - 7 | 1002 | 126 | ||
| AK104008.1 | - | 2 - 3 | 409 | 126 | ||
More Information
Set Membership: |
|

LOC4348496
Genomic Map