Assay Details
| Gene Symbol: LOC107275261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759000.2 | XP_015614486.1 | 11 - 12 | 2147 | 110 | |
| GenBank mRNA | AK062191.1 | - | 3 - 4 | 824 | 110 | |
| AK072620.1 | - | 11 - 12 | 2150 | 110 | ||
More Information
Set Membership: |
|

LOC107275261
Genomic Map