Assay Details
| Gene Symbol: LOC107275261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759000.2 | XP_015614486.1 | 3 - 4 | 452 | 107 | |
| GenBank mRNA | AK072620.1 | - | 3 - 4 | 453 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015759000.2 transcripts.
Set Membership: |
|

LOC107275261
Genomic Map