Assay Details
| Gene Symbol: LOC107275261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759000.2 | XP_015614486.1 | 6 - 7 | 1020 | 115 | |
| GenBank mRNA | AK072620.1 | - | 6 - 7 | 1023 | 115 | |
More Information
Set Membership: |
|

LOC107275261
Genomic Map