Assay Details
| Gene Symbol: LOC107275261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759000.2 | XP_015614486.1 | 9 - 10 | 1527 | 110 | |
| GenBank mRNA | AK062191.1 | - | 1 - 2 | 204 | 110 | |
| AK072620.1 | - | 9 - 10 | 1530 | 110 | ||
More Information
Set Membership: |
|

LOC107275261
Genomic Map