Assay Details
| Gene Symbol: LOC4348988 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758080.2 | XP_015613566.1 | 15 - 16 | 1273 | 90 | |
| GenBank mRNA | AK068613.1 | - | 15 - 16 | 1253 | 90 | |
More Information
Set Membership: |
|

LOC4348988
Genomic Map