Assay Details
| Gene Symbol: LOC4348988 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758079.2 | XP_015613565.1 | 1 - 2 | 258 | 132 | |
| XM_015758080.2 | XP_015613566.1 | 1 - 2 | 196 | 132 | ||
| GenBank mRNA | AB039671.1 | - | 1 - 2 | 236 | 132 | |
| AK068374.1 | - | 1 - 2 | 251 | 132 | ||
| AK068613.1 | - | 1 - 2 | 176 | 132 | ||
More Information
Set Membership: |
|

LOC4348988
Genomic Map