Assay Details
Gene Symbol: LOC4348988 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015758079.2 | XP_015613565.1 | 2 - 3 | 323 | 63 | |
XM_015758080.2 | XP_015613566.1 | 2 - 3 | 261 | 63 | ||
GenBank mRNA | AB039671.1 | - | 2 - 3 | 301 | 63 | |
AK059912.1 | - | 2 - 3 | 119 | 63 | ||
AK068374.1 | - | 2 - 3 | 316 | 63 | ||
AK068613.1 | - | 2 - 3 | 241 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
