Assay Details
| Gene Symbol: LOC4348988 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758079.2 | XP_015613565.1 | 3 - 4 | 377 | 85 | |
| XM_015758080.2 | XP_015613566.1 | 3 - 4 | 315 | 85 | ||
| XM_015758081.2 | XP_015613567.1 | 2 - 3 | 215 | 85 | ||
| GenBank mRNA | AB039671.1 | - | 3 - 4 | 355 | 85 | |
| AK059912.1 | - | 3 - 4 | 173 | 85 | ||
| AK068374.1 | - | 3 - 4 | 370 | 85 | ||
| AK068613.1 | - | 3 - 4 | 295 | 85 | ||
More Information
Set Membership: |
|

LOC4348988
Genomic Map