Assay Details
| Gene Symbol: LOC4348988 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758079.2 | XP_015613565.1 | 6 - 7 | 589 | 75 | |
| XM_015758080.2 | XP_015613566.1 | 6 - 7 | 527 | 75 | ||
| XM_015758081.2 | XP_015613567.1 | 5 - 6 | 427 | 75 | ||
| GenBank mRNA | AB039671.1 | - | 6 - 7 | 567 | 75 | |
| AF251064.1 | - | 4 - 5 | 229 | 75 | ||
| AK059912.1 | - | 6 - 7 | 385 | 75 | ||
| AK068374.1 | - | 6 - 7 | 582 | 75 | ||
| AK068613.1 | - | 6 - 7 | 507 | 75 | ||
More Information
Set Membership: |
|

LOC4348988
Genomic Map