Assay Details
| Gene Symbol: LOC4348988 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758079.2 | XP_015613565.1 | 7 - 8 | 654 | 118 | |
| XM_015758080.2 | XP_015613566.1 | 7 - 8 | 592 | 118 | ||
| XM_015758081.2 | XP_015613567.1 | 6 - 7 | 492 | 118 | ||
| GenBank mRNA | AB039671.1 | - | 7 - 8 | 632 | 118 | |
| AF251064.1 | - | 5 - 6 | 294 | 118 | ||
| AK059912.1 | - | 7 - 8 | 450 | 118 | ||
| AK068374.1 | - | 7 - 8 | 647 | 118 | ||
| AK068613.1 | - | 7 - 8 | 572 | 118 | ||
More Information
Set Membership: |
|

LOC4348988
Genomic Map