Assay Details
| Gene Symbol: LOC4348988 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758079.2 | XP_015613565.1 | 8 - 9 | 715 | 83 | |
| XM_015758080.2 | XP_015613566.1 | 8 - 9 | 653 | 83 | ||
| XM_015758081.2 | XP_015613567.1 | 7 - 8 | 553 | 83 | ||
| GenBank mRNA | AB039671.1 | - | 8 - 9 | 693 | 83 | |
| AF251064.1 | - | 6 - 7 | 355 | 83 | ||
| AK059912.1 | - | 8 - 9 | 511 | 83 | ||
| AK068374.1 | - | 8 - 9 | 708 | 83 | ||
| AK068613.1 | - | 8 - 9 | 633 | 83 | ||
More Information
Set Membership: |
|

LOC4348988
Genomic Map