Assay Details
| Gene Symbol: LOC4328238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770002.2 | XP_015625488.1 | 15 - 16 | 5660 | 59 | |
| GenBank mRNA | AK241505.1 | - | 9 - 10 | 1785 | 59 | |
More Information
Set Membership: |
|

LOC4328238
Genomic Map