Assay Details
| Gene Symbol: LOC4328443 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769351.2 | XP_015624837.1 | 2 - 3 | 346 | 68 | |
| XM_015769352.2 | XP_015624838.1 | 2 - 3 | 346 | 68 | ||
| GenBank mRNA | AK109522.1 | - | 2 - 3 | 292 | 68 | |
| DQ004266.1 | - | 2 - 3 | 280 | 68 | ||
| JX025003.1 | - | 2 - 3 | 292 | 68 | ||
More Information
Set Membership: |
|

LOC4328443
Genomic Map