Assay Details
Gene Symbol: LOC4328443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015769351.2 | XP_015624837.1 | 3 - 4 | 423 | 76 | |
XM_015769352.2 | XP_015624838.1 | 3 - 4 | 423 | 76 | ||
GenBank mRNA | AK109522.1 | - | 3 - 4 | 369 | 76 | |
DQ004266.1 | - | 3 - 4 | 357 | 76 | ||
JX025003.1 | - | 3 - 4 | 369 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
