Assay Details
| Gene Symbol: LOC4328488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770639.2 | XP_015626125.1 | 12 - 13 | 1521 | 66 | |
| GenBank mRNA | AK066421.1 | - | 7 - 8 | 734 | 66 | |
More Information
Set Membership: |
|

LOC4328488
Genomic Map