Assay Details
Gene Symbol: LOC4329236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768583.2 | XP_015624069.1 | 2 - 3 | 159 | 146 | |
XM_015768585.2 | XP_015624071.1 | 2 - 3 | 161 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
