Assay Details
| Gene Symbol: LOC4329675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770047.2 | XP_015625533.1 | 5 - 6 | 693 | 127 | |
| XM_015770048.2 | XP_015625534.1 | 5 - 6 | 687 | 127 | ||
| GenBank mRNA | AK064926.1 | - | 5 - 6 | 635 | 127 | |
More Information
Set Membership: |
|

LOC4329675
Genomic Map