Assay Details
| Gene Symbol: LOC4330983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771131.1 | XP_015626617.1 | 3 - 4 | 737 | 118 | |
| GenBank mRNA | AK068367.1 | - | 3 - 4 | 729 | 118 | |
| AK104731.1 | - | 3 - 4 | 714 | 118 | ||
| AK106174.1 | - | 3 - 4 | 714 | 118 | ||
| AK119191.1 | - | 3 - 4 | 647 | 118 | ||
More Information
Set Membership: |
|

LOC4330983
Genomic Map