Assay Details
| Gene Symbol: LOC4331357 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775508.2 | XP_015630994.1 | 7 - 8 | 874 | 102 | |
| GenBank mRNA | AB524355.1 | - | 7 - 8 | 832 | 102 | |
| AK065147.1 | - | 7 - 8 | 796 | 102 | ||
More Information
Set Membership: |
|

LOC4331357
Genomic Map