Assay Details
| Gene Symbol: LOC4332601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773710.2 | XP_015629196.1 | 9 - 10 | 1208 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4332601| Gene Symbol: LOC4332601 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773710.2 | XP_015629196.1 | 9 - 10 | 1208 | 128 | |
Set Membership: |
|