Assay Details
| Gene Symbol: LOC4332813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776553.2 | 1 - 2 | 91 | 62 | ||
| GenBank mRNA | AK073724.1 | - | 1 - 2 | 69 | 62 | |
| AK104084.1 | - | 1 - 2 | 58 | 62 | ||
| AK119262.1 | - | 1 - 2 | 65 | 62 | ||
| BT129998.1 | - | 1 - 2 | 56 | 62 | ||
| BT130563.1 | - | 1 - 2 | 66 | 62 | ||
| BT130786.1 | - | 1 - 2 | 52 | 62 | ||
| BT130973.1 | - | 1 - 2 | 82 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015776553.2 transcripts.
Set Membership: |
|

LOC4332813
Genomic Map