Assay Details
| Gene Symbol: LOC4335977 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778429.2 | XP_015633915.1 | 2 - 3 | 1073 | 77 | |
| GenBank mRNA | AB056061.1 | - | 2 - 3 | 1033 | 77 | |
| AK068340.1 | - | 2 - 3 | 1059 | 77 | ||
More Information
Set Membership: |
|

LOC4335977
Genomic Map